Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REV3L Polyclonal Antibody
REV3L Polyclonal Antibody
Supplier: Thermo Scientific PA567049
Description
REV3L Polyclonal Antibody for ICC/IF

Specifications
REV3L | |
Polyclonal | |
Unconjugated | |
REV3L | |
DNA polymerase zeta catalytic subunit; POLZ; Protein reversionless 3-like; Rev; REV3; REV3 like, DNA directed polymerase zeta catalytic subunit; Rev-3, yeast, homolog-like (polymerase, DNA, zeta); Rev3l; REV3-like; REV3-like, catalytic subunit of DNA polymerase zeta; REV3-like, catalytic subunit of DNA polymerase zeta RAD54 like (S. cerevisiae); REV3-like, polymerase (DNA directed), zeta, catalytic subunit; Seizure-related protein 4; Sez4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5980 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60673 | |
REV3L | |
Recombinant Human REV3L. Recombinant protein control fragment (Product #RP-107691). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction