Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REV3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104483 Supplier Invitrogen™ Supplier No. PA5104483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total REV3L.

Catalytic subunit of the DNA polymerase zeta complex, an error-prone polymerase specialized in translesion DNA synthesis (TLS). Lacks an intrinsic 3'-5' exonuclease activity and thus has no proofreading function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REV3L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene REV3L
Gene Accession No. O60673, Q61493
Gene Alias DNA polymerase zeta catalytic subunit; POLZ; Protein reversionless 3-like; Rev; REV3; REV3 like, DNA directed polymerase zeta catalytic subunit; Rev-3, yeast, homolog-like (polymerase, DNA, zeta); Rev3l; REV3-like; REV3-like, catalytic subunit of DNA polymerase zeta; REV3-like, catalytic subunit of DNA polymerase zeta RAD54 like (S. cerevisiae); REV3-like, polymerase (DNA directed), zeta, catalytic subunit; Seizure-related protein 4; Sez4
Gene Symbols REV3L
Host Species Rabbit
Immunogen A synthesized peptide derived from human REV3L(Accession O60673), corresponding to amino acid residues V238-D288.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19714, 5980
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.