Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REV3L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984380
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4380 Supplier Bioss Supplier No. BS19840R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Catalytic subunit of the DNA polymerase zeta complex, an error-prone polymerase specialized in translesion DNA synthesis (TLS). Lacks an intrinsic 3′-5′ exonuclease activity and thus has no proofreading function.

Specifications

Antigen REV3L
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene REV3L
Gene Accession No. O60673
Gene Alias DNA polymerase zeta catalytic subunit; POLZ; Protein reversionless 3-like; Rev; REV3; REV3 like, DNA directed polymerase zeta catalytic subunit; Rev-3, yeast, homolog-like (polymerase, DNA, zeta); REV3L; REV3-like; REV3-like, catalytic subunit of DNA polymerase zeta RAD54 like (S. cerevisiae); REV3-like, polymerase (DNA directed), zeta, catalytic subunit; Seizure-related protein 4; Sez4
Gene Symbols REV3L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human REV3L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5980
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.