Learn More
Thermo Scientific™ Reverse Osmosis Membranes
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Manufacturer: Thermo Scientific™ 09.2003
Description
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Use Membranes with the Following Barnstead Systems:
09.2003:
- Reverse osmosis membrane with integrated pretreatment, 3L/hr for Smart2Pure™ systems
09.2006:
- Reverse osmosis membrane with integrated pretreatment, 6L/hr for Smart2Pure systems
22.0046:
- LabTower™ EDI 15 requires two of these RO membranes
- LabTower™ RO 20 requires two of these RO membranes
- LabTower RO 60 requires two of these RO membranes, along with two 22.0087 RO membranes
- LabTower™ TII 20 requires two of these RO membranes
- LabTower TII 60 requires two of these RO membranes, along with two 22.0087 RO membranes
- Pacific™ RO 3, 7, 12 require one of these RO membranes
- Pacific RO 20 requires two of these RO membranes
- Pacific™ TII 3, 7, 12 require one of these RO membranes
- Pacific TII 20 requires two of these RO membranes
22.0087:
- LabTower™ EDI 30 requires two of these RO membranes
- LabTower™ RO 40 requires two of these RO membranes
- LabTower RO 60 requires two of these RO membranes, along with two 22.0046 RO membranes
- LabTower TII 40 requires two of these RO membranes
- LabTower TII 60 requires two of these RO membranes, along with two 22.0046 RO membranes
- Pacific RO 40 requires two of these RO membranes
- Pacific TII 40 requires two of these RO membranes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Reverse Osmosis membrane with integrated pretreatment, 3L/hr | |
Smart2Pure | |
Reverse Osmosis Membrane (pretreated) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.