Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Reverse Transcriptase (HIV) Recombinant Protein

Catalog No. 89959782
Click to view available options
Quantity:
200 U

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Formulation In 10mM potassium phosphate, pH 7.4, 1mM DTT and 20% glycerol
Name Reverse Transcriptase (HIV) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 200 U
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name REVERSE
Assays Reverse Transcriptase Assay; AIDS research; Transcribe mRNA into double stranded cDNA; Used with either single stranded DNA or RNA templates to make probes for use in hybridization experiments; Labeling the termini of DNA fragments with protruding 5' termini.
Biological Activity ≥10,000 units per mg protein. One Unit incorporates 1nmole of tritiated d-TMP into acid precipitable products using poly(A)/oligo(dT)12-18 as the template/primer in 20 minutes at 37°C, pH 8.3.
Species E. coli
Protein Tag None
Expression System Escherichia coli expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.