Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience Revogene™ SARS-CoV-2

Catalog No. 23029057 Shop All Meridian Bioscience Products
Change view
Click to view available options
Detectable Analytes:
SARS-CoV-2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-029-057 SARS-CoV-2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-057 Supplier Meridian Bioscience Supplier No. 410700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Detects SARS-CoV-2 RNA from nasopharyngeal, oropharyngeal, anterior nasal, and mid-turbinate swab samples.

  • The Revogene™ SARS-CoV-2 assay is a real-time RT-PCR test intended for the qualitative detection of RNA from SARS-CoV-2 in nasopharyngeal, oropharyngeal, anterior nasal, and mid-turbinate nasal swab specimens from individuals suspected of COVID-19 by their healthcare provider
  • Testing is for laboratories certified under the Clinical Laboratory Improvement Amendments of 1988 (CLIA), 42 U.S.C. § 263a, that meet requirements to perform moderate or high complexity tests
  • The Revogene™ platform features a fully automated molecular workflow designed to optimize COVID-19 testing capacity
  • With its intuitive, press-and-walk-away design, the system streamlines operations into just two easy steps and requires less than one minute of hands-on time
  • The assay can deliver positive results as early as 47 min, with negative results available in 85 min

Specifications

CE Marker No
Clia Complexity Moderate
Detectable Analytes SARS-CoV-2
Detection Method RT-PCR
Sample Type Nasopharyngeal, Oropharyngeal. Anterior Nasal, Mid-Turbinate Nasal Swab Sample
Type COVID Test Kit
Disposable Single-use
For Use With (Application) Nasopharyngeal, Oropharyngeal, Anterior Nasal, and Mid-Turbinate Nasal Swab Specimens
For Use With (Equipment) Revogene Instrument (23-029-051)
High-throughput Compatibility 8 Samples/Run
Includes 24 Disposable Transfer Tools, 24 Individual Pouches, Each Containing Revogene SARS CoV-2 Microfluidic Cartridge (PIE)
Quantity 24 Tests/kit
Sterility Sterile
Analysis Time 47 min. early call feature/85 min. result
Form Ready-to-Use (RTU)
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.