Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rex-1/ZFP42 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299539PCP
Description
Rex-1/ZFP42 Polyclonal antibody specifically detects Rex-1/ZFP42 in Mouse samples. It is validated for Western BlotSpecifications
Rex-1/ZFP42 | |
Polyclonal | |
Western Blot | |
FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila) | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse Rex-1/ZFP42. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
132625 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction