Learn More
Invitrogen™ Rex1 Monoclonal Antibody (5E11A6)

Description
This antibody has been tested in indirect ELISA.
Specifications
Specifications
| Antigen | Rex1 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 5E11A6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | ZFP42 |
| Gene Accession No. | P22227, Q96MM3 |
| Gene Alias | 1700021P10Rik; 2610511M11Rik; AW050347; ELOABP1; EloA-BP1; elongin A binding protein 1; elongin A-binding protein 1; elongin-A-binding protein 1; hREX-1; KIAA1138; mREX-1; reduced expression protein 1; REX1; Rex-1; REX1 transcription factor; REX1, RNA exonuclease 1 homolog; REX1, RNA exonuclease 1 homolog (S. cerevisiae); REXO1; RNA exonuclease 1 homolog; Tceb3bp1; transcription elongation factor B polypeptide 3 binding protein 1; transcription elongation factor B polypeptide 3-binding protein 1; Zfp42; Zfp-42; ZFP42 zinc finger protein; Zfp971; Zinc finger protein 42; zinc finger protein 42 homolog; Zinc finger protein 754; zinc finger protein 971; ZNF754 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.