Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rex1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585867 Supplier Invitrogen™ Supplier No. PA585867
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rex1 Polyclonal Antibody for Western Blot

Involved in the reprogramming of X-chromosome inactivation during the acquisition of pluripotency. Required for efficient elongation of TSIX, a non-coding RNA antisense to XIST. Binds DXPas34 enhancer within the TSIX promoter. Involved in ES cell self-renewal. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rex1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene ZFP42
Gene Accession No. Q96MM3
Gene Alias 1700021P10Rik; 2610511M11Rik; AW050347; ELOABP1; EloA-BP1; elongin A binding protein 1; elongin A-binding protein 1; elongin-A-binding protein 1; hREX-1; KIAA1138; mREX-1; reduced expression protein 1; REX1; Rex-1; REX1 transcription factor; REX1, RNA exonuclease 1 homolog; REX1, RNA exonuclease 1 homolog (S. cerevisiae); REXO1; RNA exonuclease 1 homolog; Tceb3bp1; transcription elongation factor B polypeptide 3 binding protein 1; transcription elongation factor B polypeptide 3-binding protein 1; Zfp42; Zfp-42; ZFP42 zinc finger protein; Zfp971; Zinc finger protein 42; zinc finger protein 42 homolog; Zinc finger protein 754; zinc finger protein 971; ZNF754
Gene Symbols ZFP42
Host Species Rabbit
Immunogen Synthetic peptide within Human ZFP-42/REX-1 aa 261-310.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132625
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.