Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REXO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559894 Supplier Invitrogen™ Supplier No. PA559894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSGKYVVDNS RPPTDLEYDP LSNYSARHLS RASSRDERAA KRPRGSRGSE PYTPAPKKLC DPFGSCDARF SDSEDEAATV PGNEPTT Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 70%.

REXO1, also named as EloA-BP1, ELOABP1, KIAA1138 and TCEB3BP1, is transcription elongation factor B polypeptide 3-binding protein 1. REXO1 did not affect either the rate of transcription elongation by RNA polymerase II or the transcriptional activity of TCEB3 in vitro. This gene is different to the REX1 which is an embryo stem cell marker.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REXO1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene REXO1
Gene Accession No. Q8N1G1
Gene Alias 1700021P10Rik; 2610511M11Rik; AW050347; ELOABP1; EloA-BP1; elongin A binding protein 1; elongin A-binding protein 1; elongin-A-binding protein 1; Kiaa1138; REX1; REX1, RNA exonuclease 1 homolog; REX1, RNA exonuclease 1 homolog (S. cerevisiae); Rexo1; RNA exonuclease 1 homolog; TCEB3BP1; transcription elongation factor B polypeptide 3 binding protein 1; transcription elongation factor B polypeptide 3-binding protein 1
Gene Symbols REXO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human REXO1. Recombinant protein control fragment (Product #RP-98888).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.