Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REXO1 Polyclonal Antibody

REXO1 Polyclonal Antibody
Supplier: Thermo Scientific PA5111157
Description
REXO1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| REXO1 | |
| Polyclonal | |
| Unconjugated | |
| REXO1 | |
| 1700021P10Rik; 2610511M11Rik; AW050347; ELOABP1; EloA-BP1; elongin A binding protein 1; elongin A-binding protein 1; elongin-A-binding protein 1; Kiaa1138; REX1; REX1, RNA exonuclease 1 homolog; REX1, RNA exonuclease 1 homolog (S. cerevisiae); Rexo1; RNA exonuclease 1 homolog; TCEB3BP1; transcription elongation factor B polypeptide 3 binding protein 1; transcription elongation factor B polypeptide 3-binding protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57455 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.60 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N1G1 | |
| REXO1 | |
| Recombinant Protein corresonding to Human REXO1. Recombinant protein control fragment (Product #RP-98887). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction