Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REXO2 Antibody (59D9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242639FR
Description
REXO2 Monoclonal specifically detects REXO2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
REXO2 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
REXO2 | |
Recombinant human REXO 2 (26-237aa) purified from E. coli | |
100 μL | |
DNA replication Transcription Translation and Splicing | |
25996 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
59D9 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
CGI-114, DKFZP566E144, EC 3.1, oligoribonuclease, mitochondrial, REX2, REX2, RNA exonuclease 2 homolog (S. cerevisiae), RFN, RNA exonuclease 2 homolog, SFNMGC111570, Small fragment nuclease, SMFN | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction