Learn More
REXO2 Monoclonal Antibody (2B5)

Description
REXO2 Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | REXO2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2B5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | REXO2 |
| Gene Accession No. | Q9Y3B8 |
| Gene Alias | 1810038D15Rik; AW107347; C11orf61; CGI-114; Gm13138; novel KRAB box domain containing protein; oligoribonuclease, mitochondrial; OTTMUSG00000010655; reduced expression 2; REX2; REX2, RNA exonuclease 2 homolog; REX2, RNA exonuclease 2 homolog (S. cerevisiae); Rexo2; RFN; RNA exonuclease 2; RNA exonuclease 2 homolog; SFN; small fragment nuclease; SMFN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.