Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ REXO2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | REXO2 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119403
|
Invitrogen™
PA5119403 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5121644
|
Invitrogen™
PA5121644 |
20 μL |
Each for $237.00
|
|
|||||
Description
REXO2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Nucleases are components of DNA and RNA metabolism that carry out functions in DNA repair, replication, and recombination and in RNA processing and degradation. SFN is a homolog of Orn, a 3-prime-to-5-prime exoribonuclease of E. coli that attacks the free 3-prime hydroxyl group on single-stranded RNA, releasing 5-prime mononucleotides in a sequential manner.Specifications
REXO2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
REXO2 | |
1810038D15Rik; AW107347; C11orf61; CGI-114; Gm13138; novel KRAB box domain containing protein; oligoribonuclease, mitochondrial; OTTMUSG00000010655; reduced expression 2; REX2; REX2, RNA exonuclease 2 homolog; REX2, RNA exonuclease 2 homolog (S. cerevisiae); Rexo2; RFN; RNA exonuclease 2; RNA exonuclease 2 homolog; SFN; small fragment nuclease; SMFN | |
REXO2 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q9Y3B8 | |
25996 | |
Recombinant protein RNA Exonuclease 2 Homolog. The antigen corresponds to amino acid range 41-213 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title