Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REXO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590722 Supplier Invitrogen™ Supplier No. PA590722
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VREGGAAMAA GESMAQRMVW VDLEMTGLDI EKDQIIEMAC LITDSDLNIL AEGPNLIIKQ PDELLDSMSD WCKEHHGKSG LTKAVKESTI TLQQAEYEFL SFVRQQTPPG LCPLAGNSVH EDKKFLDKYM PQFMKHLHYR IIDVSTVKEL CRRWYPEEYE FAPKKAASHR ALDDISESIK ELQFYRNNIF KKKIDEKKRK IIENGENEKT VS; Positive Samples: 293T, U-87MG, LO2, HT-29; Cellular Location: Cytoplasm, Mitochondrion intermembrane space, Mitochondrion matrix, Nucleus.

Nucleases are components of DNA and RNA metabolism that carry out functions in DNA repair, replication, and recombination and in RNA processing and degradation. SFN is a homolog of Orn, a 3-prime-to-5-prime exoribonuclease of E. coli that attacks the free 3-prime hydroxyl group on single-stranded RNA, releasing 5-prime mononucleotides in a sequential manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REXO2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene REXO2
Gene Accession No. Q5U1X1, Q9D8S4, Q9Y3B8
Gene Alias 1810038D15Rik; AW107347; C11orf61; CGI-114; Gm13138; novel KRAB box domain containing protein; oligoribonuclease, mitochondrial; OTTMUSG00000010655; reduced expression 2; REX2; REX2, RNA exonuclease 2 homolog; REX2, RNA exonuclease 2 homolog (S. cerevisiae); Rexo2; RFN; RNA exonuclease 2; RNA exonuclease 2 homolog; SFN; small fragment nuclease; SMFN
Gene Symbols Rex2, REXO2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 26-237 of human REXO2 (NP_0563382).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100043034, 100362123, 104444, 25996, 300689
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.