Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REXO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561399 Supplier Invitrogen™ Supplier No. PA561399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEWLLKQKSQ APEKPLVISQ MGSKKKPKII QQNKKETSPQ VKGEEMPAGK DQEASRGSVP SGSKMDRRAP VPRTKASGTE HNKKGTKERT NGDIVPERGD IEHKKRKAKE AAPAPPTEED IWFDDVDP Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 52%.

REXO4 (REX4 Homolog, 3'-5' Exonuclease) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includenucleic acid bindingandDNA-binding transcription factor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REXO4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene REXO4
Gene Accession No. Q9GZR2
Gene Alias exonuclease hPMC2; exonuclease XPMC2; Gm111; hPMC2; Pmc2; Prevents mitotic catastrophe 2 protein homolog; REX4; REX4 homolog, 3'-5' exonuclease; REX4, RNA exonuclease 4 homolog; REX4, RNA exonuclease 4 homolog (S. cerevisiae); REXO4; RNA exonuclease 4; Xenopus prevents mitotic catastrophe 2 homolog; XPMC2; XPMC2 prevents mitotic catastrophe 2 homolog; XPMC2H
Gene Symbols REXO4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human REXO4. Recombinant protein control fragment (Product #RP-89359).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57109
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.