Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REXO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589811 Supplier Invitrogen™ Supplier No. PA589811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WKALQEWLLK QKSQAPEKPL VISQMGSKKK PKIIQQNKKE TSPQVKGEEM PAGKDQEASR GSVPSGSKMD RRAPVPRTKA SGTEHNKKGT KERTNGDIVP ERGDIEHKKR KAKEAAPAPP TEEDIWFDDV DPADIEAAIG PEAAKIARKQ LGQSEGSVSL SLVKEQAFGG L; Positive Samples: HeLa, LO2, Jurkat, HepG2, U-87MG; Cellular Location: Nucleus, nucleolus.

REXO4 (REX4 Homolog, 3'-5' Exonuclease) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includenucleic acid bindingandDNA-binding transcription factor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REXO4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene REXO4
Gene Accession No. Q9GZR2
Gene Alias exonuclease hPMC2; exonuclease XPMC2; Gm111; hPMC2; Pmc2; Prevents mitotic catastrophe 2 protein homolog; REX4; REX4 homolog, 3'-5' exonuclease; REX4, RNA exonuclease 4 homolog; REX4, RNA exonuclease 4 homolog (S. cerevisiae); REXO4; RNA exonuclease 4; Xenopus prevents mitotic catastrophe 2 homolog; XPMC2; XPMC2 prevents mitotic catastrophe 2 homolog; XPMC2H
Gene Symbols REXO4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 70-240 of human REXO4 (NP_0651182).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57109
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.