Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFC1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB243050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB243050 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB243050 Supplier Novus Biologicals Supplier No. NB100230G
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RFC1 Polyclonal antibody specifically detects RFC1 in Human samples. It is validated for Western Blot

Specifications

Antigen RFC1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias A1, A1 140 kDa subunit, Activator 1 140 kDa subunit, Activator 1 large subunit, Activator 1 subunit 1, MGC51786, MHC binding factor, beta, MHCBFB, PO-GA, RECC1, replication factor C (activator 1) 1 (145kD), replication factor C (activator 1) 1, 145kDa, Replication factor C 140 kDa subunit, Replication factor C large subunit, replication factor C subunit 1, replication factor C1, RFC, RF-C 140 kDa subunit, RFC140DNA-binding protein PO-GA
Host Species Goat
Immunogen The immunogen recognized by this antibody maps to a region between residue 1100 and the C-terminus (residue 1148) of human Replication Factor C subunit 1 using the numbering given in entry NP_002904.3 (GeneID 5981).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5981
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.