Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFC1 Polyclonal Antibody

Catalog No. PIPA535965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535965 Supplier Invitrogen™ Supplier No. PA535965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (88%), Rat (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Replication factor C (RFC) is an essential DNA polymerase accessory protein that is required for numerous aspects of DNA metabolism, including DNA replication, DNA repair and telomere metabolism. RFC is a heteropentameric complex that recognizes a primer on a template DNA, binds to a primer terminus and loads proliferating cell nuclear antigen (PCNA) onto DNA at primer-template junctions in an ATP-dependent reaction. All five of the RFC subunits share a set of related sequences (RFC boxes) that include nucleotide binding consensus sequences. Four of the five RFC genes (RFC1, RFC2, RFC3 and RFC4) have consensus ATP-binding motifs. The small RFC proteins, RFC2, RFC3, RFC4 and RFC5, interact with Rad24, whereas the RFC1 subunit does not. RFC2, the third-largest subunit of the RFC complex, exhibits ATP binding which makes it important for both DNA replication and checkpoint function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RFC1
Gene Accession No. P35251
Gene Alias 140kDa; A1; A1 140 kDa subunit; A1-P145; Activator 1 140 kDa subunit; activator 1 large subunit; Activator 1 subunit 1; Alp145; Differentiation-specific element-binding protein; DNA-binding protein PO-GA; Ibf-1; ISRE-binding protein; MHC binding factor, beta; MHCBFB; PO-GA; RECC1; replication factor C (activator 1) 1; replication factor C (activator 1) 1, 145kDa; replication factor C 1; replication factor C 140 kDa subunit; Replication factor C large subunit; replication factor C subunit 1; Replication factor C subunit 1-like protein; replication factor C, 140 kDa; replication factor C1; RFC; RF-C 140 kDa subunit; Rfc1; RFC140
Gene Symbols RFC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human RFC1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5981
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.