Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFC2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100231AF488
Description
RFC2 Polyclonal antibody specifically detects RFC2 in Human samples. It is validated for Western BlotSpecifications
RFC2 | |
Polyclonal | |
Western Blot | |
A1, A1 40 kDa subunit, Activator 1 40 kDa subunit, Activator 1 subunit 2, MGC3665, replication factor C (activator 1) 2, 40kDa, replication factor C 2, Replication factor C 40 kDa subunit, replication factor C subunit 2, RF-C 40 kDa subunit, RFC40replication factor C (activator 1) 2 (40kD) | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Replication Factor C subunit 2 using the numbering given in entry NP_852136.1 (GeneID 5982). | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair | |
5982 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction