Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFC2 Antibody (OTI4C11), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273858JF549
Description
RFC2 Monoclonal specifically detects RFC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
RFC2 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RFC2 | |
Recombinant protein fragment corresponding to amino acids 1-234 of human RFC2 (NP_002905) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4C11 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
A1, A1 40 kDa subunit, Activator 1 40 kDa subunit, Activator 1 subunit 2, MGC3665, replication factor C (activator 1) 2, 40kDa, replication factor C 2, Replication factor C 40 kDa subunit, replication factor C subunit 2, RF-C 40 kDa subunit, RFC40replication factor C (activator 1) 2 (40kD) | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, DNA Repair | |
5982.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction