Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFC3 Monoclonal Antibody (OTI1C7), OriGene
SDP

Catalog No. 502543880
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3880 Supplier OriGene Supplier No. CF811943
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RFC3 Monoclonal antibody specifically detects RFC3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RFC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C7
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CELF6
Gene Accession No. Q96J87
Gene Alias 2810416I22Rik; 38kDa; A1 38 kDa subunit; Activator 1 38 kDa subunit; activator 1 subunit 3; AU022547; DNA-binding; MGC5276; Polymorphism; Recc3; replication factor C (activator 1) 3; replication factor C (activator 1) 3, 38kDa; replication factor C 3; replication factor C 38 kDa subunit; Replication factor C subunit 3; replication factor C, 38 kDa; RF-C 38 kDa subunit; RFC, 38 kD subunit; RFC3; RFC38
Host Species Mouse
Immunogen Full length human recombinant protein of human CELF6 (NP_443072) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60677
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.