Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFC3 Monoclonal Antibody (OTI4A7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811875

Description
The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa. This gene encodes the 38 kDa subunit. This subunit is essential for the interaction between the 140 kDa subunit and the core complex that consists of the 36, 37, and 40 kDa subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
RFC3 | |
Monoclonal | |
Unconjugated | |
RFC3 | |
RFC38 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5983 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI4A7 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P40938 | |
RFC3 | |
Full length human recombit protein of human RFC3 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction