Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFC4 Antibody, DyLight 650, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100233C
Description
RFC4 Polyclonal antibody specifically detects RFC4 in Human, Mouse samples. It is validated for Western BlotSpecifications
RFC4 | |
Polyclonal | |
Western Blot | |
A1, A1 37 kDa subunit, Activator 1 37 kDa subunit, Activator 1 subunit 4, MGC27291, replication factor C (activator 1) 4, 37kDa, Replication factor C 37 kDa subunit, replication factor C subunit 4, RFC 37 kDa subunit, RF-C 37 kDa subunit, RFC37replication factor C (activator 1) 4 (37kD) | |
The immunogen recognized by this antibody maps to a region between residues 300 and 350 of human Replication Factor C subunit 4 using the numbering given in entry NP_002907.1 (GeneID 5984). | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair | |
5984 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction