Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFC4 Antibody (OTI1A8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273859H
Description
RFC4 Monoclonal specifically detects RFC4 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RFC4 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
RFC4 | |
Full length recombinant protein of human RFC4 (NP_002907) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1A8 | |
Western Blot, Immunohistochemistry | |
A1, A1 37 kDa subunit, Activator 1 37 kDa subunit, Activator 1 subunit 4, MGC27291, replication factor C (activator 1) 4, 37kDa, Replication factor C 37 kDa subunit, replication factor C subunit 4, RFC 37 kDa subunit, RF-C 37 kDa subunit, RFC37replication factor C (activator 1) 4 (37kD) | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, DNA Repair | |
5984.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction