Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583954 Supplier Invitrogen™ Supplier No. PA583954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RIIEPLTSRC SKFRFKPLSD KIQQQRLLDI AKKENVKISD EGIAYLVKVS EGDLRKAITF LQSATRLTGG KEITEKVITD IAGVIP Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 91%.

Human Replication Factor C (RFC) is a hetero-pentameric protein consisting of the p140, p40, p38, p37 and p36 subunits. RFC functions in the loading of the DNA polymerase delta processivity factor (PCNA, Proliferating Cell nuclear Antigen) onto a primed DNA template during DNA synthesis and repair. RFC may also participate in the DNA damage checkpoint response with a PCNA-like hetero-trimer consisting of Hus 1, Rad 1 and Rad 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFC4
Gene Accession No. P35249
Gene Alias A1; A1 37 kDa subunit; Activator 1 37 kDa subunit; activator 1 subunit 4; AI894123; AU040575; EGK_12194; replication factor C (activator 1) 4; replication factor C (activator 1) 4, 37kDa; replication factor C 37 kDa subunit; Replication factor C subunit 4; RFC 37 kDa subunit; RF-C 37 kDa subunit; RFC p37; RFC37; RFC4
Gene Symbols RFC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RFC4. Recombinant protein control fragment (Product #RP-104105).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.