Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFC5 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100235AF700
Description
RFC5 Polyclonal antibody specifically detects RFC5 in Human samples. It is validated for Western BlotSpecifications
RFC5 | |
Polyclonal | |
Western Blot | |
Activator 1 36 kDa subunit, Activator 1 subunit 5,36.5 kD subunit, MGC1155, replication factor C (activator 1) 5, 36.5kDa, RF-C 36 kDa subunit, RFC36replication factor C (activator 1) 5 (36.5kD) | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Replication Factor C subunit 5 using the numbering given in entry NP_031396.1 (GeneID 5985). | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair | |
5985 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction