Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596912 Supplier Invitrogen™ Supplier No. PA596912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LMVPRLEHVV EEEKVDISED GMKALVTLSS GDMRRALNIL QSTNMAFGKV TEETVYTCTG HPLKSDIANI LDWMLNQDFT TAYRNITELK TLKGLALHDI LTEIHLFVHR VDFPSSVRIH LLTKMADIEY RLSVGTNEKI QLSSLIAAFQ VTRDLIVAEA; Positive Samples: HeLa, U-251MG, SKOV3, MCF-7, Mouse liver, Mouse spleen; Cellular Location: Nucleus.

The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kD. This gene encodes the 36 kD subunit. This subunit can interact with the C-terminal region of PCNA. It forms a core complex with the 38 and 40 kDa subunits. The core complex possesses DNA-dependent ATPase activity, which was found to be stimulated by PCNA in an in vitro system. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RFC5
Gene Accession No. P40937, Q9D0F6
Gene Alias 2610020K06Rik; 2610209F07Rik; 36.5kDa; 36kDa; A1 36 kDa subunit; activator 1 36 kDa subunit; Activator 1 subunit 5; MGC1155; Recc5; replication factor C (activator 1) 5; replication factor C (activator 1) 5, 36.5kDa; replication factor C 36 kDa subunit; replication factor C 5; replication factor C subunit 5; replication factor c subunit 5-like protein; replication factor C subunit RFC5; replication factor C, 36 kDa; replication factor C, 36-kDa subunit; RF-C 36 kDa subunit; RFC, 36.5 kD subunit; RFC36; Rfc5; rfc5 protein; Unknown (protein for MGC:128229); zgc:110313
Gene Symbols RFC5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 181-340 of human RFC5 (NP_0313961).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304528, 5985, 72151
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.