Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFESD Antibody (OTI2A7), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273860AF532
Description
RFESD Monoclonal specifically detects RFESD in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RFESD | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
RFESD | |
Full length recombinant protein of human RFESD (NP_775498) produced in HEK293T cells. | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2A7 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Rieske (Fe-S) domain containing, Rieske domain-containing protein | |
Mouse | |
0.1 mL | |
317671 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction