Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFESD Antibody (OTI2A7) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273860
Description
RFESD Monoclonal specifically detects RFESD in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RFESD | |
Monoclonal | |
LYOPH | |
Western Blot 1:4000, Immunocytochemistry/Immunofluorescence 1:100 | |
Rieske (Fe-S) domain containing, Rieske domain-containing protein | |
Mouse | |
17.6 kDa | |
100 μg | |
317671 | |
Human, Mouse | |
Lyophilized |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2A7 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
RFESD | |
Full length human recombinant protein of human RFESD(NP_775498) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction