Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFESD Antibody (OTI2A7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273860F
Description
RFESD Monoclonal specifically detects RFESD in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RFESD | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
RFESD | |
Full length recombinant protein of human RFESD (NP_775498) produced in HEK293T cells. | |
0.1 mL | |
317671.0 | |
Store at 4°C in the dark. |
Immunocytochemistry, Immunofluorescence | |
OTI2A7 | |
Immunocytochemistry/Immunofluorescence | |
Rieske (Fe-S) domain containing, Rieske domain-containing protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction