Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFFL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553666 Supplier Invitrogen™ Supplier No. PA553666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQATSVPPAQ VQENQQANGH VSQDQEEPVY LESVARVPAE DETQSIDSED SFVPGRRASL SDLTDLEDIE GLTVRQLKEI LARNF Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

Has E3 ubiquitin protein ligase activity. Regulates the levels of CASP8 and CASP10 by targeting them for proteasomal degradation. Has anti-apoptotic activity. May bind phosphatidylinositol phosphates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFFL
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rffl
Gene Accession No. Q8WZ73
Gene Alias 1700051E09Rik; 4930516L10Rik; BG080975; CARP2; CARP-2; caspase regulator CARP2; Caspases-8 and -10-associated RING finger protein 2; E3 ubiquitin-protein ligase rififylin; Fring; FYVE-RING finger protein SAKURA; RFFL; RIFIFYLIN; ring finger and FYVE like domain containing E3 ubiquitin protein ligase; ring finger and FYVE like domain containing protein; ring finger and FYVE-like domain containing 1; ring finger and FYVE-like domain containing E3 ubiquitin protein ligase; RING finger and FYVE-like domain-containing protein 1; RING finger protein 189; RING finger protein 34-like; RING finger protein SAKURA; RING-type E3 ubiquitin transferase rififylin; RNF189; RNF34L
Gene Symbols Rffl
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RFFL. Recombinant protein control fragment (Product #RP-92092).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117584
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.