Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFK Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300208AF405
Description
RFK Polyclonal specifically detects RFK in Human samples. It is validated for ELISA.Specifications
RFK | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 405 | |
ATP:riboflavin 5'-phosphotransferase, EC 2.7.1.26, Flavokinase | |
Produced in rabbits immunized with purified, recombinant RFK (Q969G6; Met1-His155). | |
0.1 mL | |
Lipid and Metabolism | |
55312 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction