Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFK Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551222

Catalog No. PA551222


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total RFK protein.

Riboflavin kinase is an essential enzyme that catalyzes the phosphorylation of riboflavin to form flavin mononucleotide, an obligatory step in vitamin B2 utilization and flavin cofactor synthesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RFK
Polyclonal
Unconjugated
RFK
0610038L10Rik; AF031381; ATP:riboflavin 5'-phosphotransferase; flavokinase; KOI-4; Rfk; Riboflavin kinase; RIFK
Rabbit
Antigen affinity chromatography
RUO
55312
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q969G6
RFK
Full length fusion protein of human RFK.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.