Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFLAT-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100594 Supplier Invitrogen™ Supplier No. PA5100594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KLF13.

RANTES factor of late activated T-lymphocytes 1 (RFAT-1) represses transcription by binding to the BTE site, a GC-rich DNA element, in competition with the activator SP1. It also represses transcription by interacting with the corepressor Sin3A and HDAC1. Activates RANTES expression in T-cells. (Uniprot).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFLAT-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KLF13
Gene Accession No. Q9Y2Y9
Gene Alias 0610043C13Rik; 9430029L20Rik; basic transcription element binding protein 3; basic transcription element-binding protein 3; Bteb3; BTE-binding protein 3; C2-H2 zinc finger protein; erythroid transcription factor FKLF-2; FKLF2; FKLF-2; KLF13; Krueppel-like factor 13; Kruppel like factor 13; Kruppel-like factor 13; novel Sp1-like zinc finger transcription factor 1; NSLP1; RANTES factor of late activated T lymphocytes-1; RANTES factor of late activated T-lymphocytes 1; RFLAT1; RFLAT-1; RGD1565099; Transcription factor BTEB3; transcription factor NSLP1
Gene Symbols KLF13
Host Species Rabbit
Immunogen A synthesized peptide derived from human KLF13(Accession Q9Y2Y9), corresponding to amino acid residues A250-P288.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499171, 51621
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.