Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFNG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565105
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565105 Supplier Invitrogen™ Supplier No. PA565105
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLRPDDVFIAV KTTRKNHGPR LRLLLRTWIS RARQQTFIFT DGDDPELELQ GGDRVINTNC SAVRTRQALC CKMSVEYDKF IESGRKWFCH VDDDNYVNAR SLLHLLSSFS PSQDVYLGRP SLDHPIEATE RV Highest antigen sequence identity to the following orthologs - mouse 87%, rat 88%.

Glycosyltransferase that initiates the elongation of O- linked fucose residues attached to EGF-like repeats in the extracellular domain of Notch molecules. May be involved in limb formation and in neurogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFNG
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFNG
Gene Accession No. Q9Y644
Gene Alias Beta-1,3-N-acetylglucosaminyltransferase radical fringe; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; radical fringe gene homolog; radical fringe homolog; Rfng; RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Symbols RFNG
Host Species Rabbit
Immunogen Recombinant Human RFNG. Recombinant protein control fragment (Product #RP-105861).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.