Learn More
RFP Antibody (1433), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP243580
Catalog No. NBP243580
Only null left
Add to Cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunoprecipitation
RFP Monoclonal specifically detects RFP in Non-species specific samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
RFP
Monoclonal
Unconjugated
PBS with No Preservative
Mouse
27 kDa
0.1 mL
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunoprecipitation
1433
Western Blot 1:5000-1:20000, Immunoprecipitation 1:100-1:500
DSRED, GFP-like chromoprotein, Red Fluorescent Protein, RFP
Recombinant protein conjugated synthetic peptide encompassing a sequence within the center region of RFP. The exact sequence is proprietary.
Protein G purified
RUO
Non-species specific
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only