Learn More
RFP Monoclonal Antibody (8E5.G7), Rockland™

Mouse Monoclonal Antibody
$174.73 - $619.37
Specifications
Antigen | RFP |
---|---|
Clone | 8E5.G7 |
Concentration | 1.02 mg/mL |
Applications | ELISA, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-105-7544
![]() ![]() |
Rockland Immunochemicals
200301379 |
100 μg |
Each for $619.37
|
|
|||||
50-105-7545
![]() ![]() |
Rockland Immunochemicals
200301379S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Expect reactivity against RFP and its variants: mCherry, tdTomato, mBanana, mOrange, mPlum, mOrange and mStrawberry.
Red Fluorescent Protein (RPF) is isolated from sea anemone (Discosoma sp), and commonly used as a molecular tag to study protein localization, reporter gene expression, and protein-protein interactions. Although it exists as a homotetramer, monomeric variants have been constructed for use in various applications.Specifications
RFP | |
1.02 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
RFP tag | |
IgG2a κ | |
-20°C, Avoid Freeze/Thaw Cycles |
8E5.G7 | |
ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Anti-RFP monoclonal antibody is a Red Fluorescent Protein (RFP) fusion protein corresponding to the full length amino acid sequence (234aa) derived from the mushroom polyp coral Discosoma. | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.