Learn More
Invitrogen™ RFP Recombinant Superclonal™ Antibody
Rabbit Recombinant Superclonal Antibody
Supplier: Invitrogen™ 710530
Description
Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
Red Fluorescent Protein (RFP) is isolated from sea anemone (Discosoma sp, Uniprot ID: Q9U6Y8), and commonly used as a molecular tag to study protein localization, reporter gene expression, and protein-protein interactions. Although it exists as a homotetramer, monomeric variants have been constructed for use in various applications.
Specifications
RFP | |
Recombinant Superclonal | |
Unconjugated | |
DsRed2; mRFP; red fluorescent; Red fluorescent protein; RFP; RFP tag | |
Peptide corresponding to amino acids 32-45 of Red Fluorescent Protein. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Rabbit | |
Protein A | |
RUO | |
Tag | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.