Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFPL1 Antibody (OTI1D3), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273861FR
Description
RFPL1 Monoclonal specifically detects RFPL1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
RFPL1 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
RFPL1 | |
Full length recombinant protein of human RFPL1 (NP_066306) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1D3 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
MGC132428, ret finger protein-like 1, RFPL1L, RNF78RING finger protein 78 | |
Mouse | |
Affinity Purified | |
Zinc Finger | |
5988.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction