Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFPL1 Antibody (OTI1D3), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273861MFV450
Description
RFPL1 Monoclonal antibody specifically detects RFPL1 in Human samples. It is validated for Flow CytometrySpecifications
RFPL1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI1D3 | |
Flow Cytometry | |
MGC132428, ret finger protein-like 1, RFPL1L, RNF78RING finger protein 78 | |
Full length human recombinant protein of human RFPL1 (NP_066306) produced in HEK293T cell. | |
0.1 mL | |
Zinc Finger | |
5988 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction