Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RFPL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571359
Description
RFPL1 Polyclonal Antibody for Western Blot
RFPL1 is a protein coding gene, and negatively regulates the G2-M phase transition, possibly by promoting cyclin B1/CCNB1 and CDK1 proteasomal degradation and thereby preventing their accumulation during interphase.
Specifications
RFPL1 | |
Polyclonal | |
Unconjugated | |
RFPL1 | |
ret finger protein like 1; ret finger protein-like 1; RFPL1; RFPL1L; RING finger protein 78; RNF78 | |
Rabbit | |
Affinity chromatography | |
RUO | |
5988 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75677 | |
RFPL1 | |
synthetic peptide directed towards the C terminal region of human RFPL1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction