Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFPL2/3 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001169 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001169 Supplier Novus Biologicals Supplier No. NB1001169
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RFPL2/3 Polyclonal specifically detects RFPL2/3 in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen RFPL2/3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006596
Gene Alias ret finger protein-like 2, RNF79RING finger protein 79
Gene Symbols RFPL2
Host Species Goat
Immunogen Peptide with sequence C-TTDAPVRPGEAK corresponding to C-Terminus according to NP_006596.2, NP_001091997.2, NP_001153017.1, NP_001153018.1, NP_001092005.1, NP_006595.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10739
Test Specificity This is expected to recognize both RFPL2 (NP_006596.2; NP_001091997.2; NP_001153017.1; NP_001153018.1) and RFPL3 (NP_001092005.1; NP_006595.1), which are virtually identical. Variants (NP_001153017.1; NP_001153018.1) encode the same isoform.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.