Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577249
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577249 Supplier Invitrogen™ Supplier No. PA577249
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

RFPL1, RFPL2 and RFPL3 (ret finger protein-like 1, 2 and 3, respectively), exist as a cluster of genes mapping to human chromosome 22q12.2-q13.3, sharing 95%-96% identity. RFPL1, 2 and 3, are thought to contribute to neocortex organization and size in primates, and show high expression in fetal neocortex as well as embryonic stem-cell neurogenesis. Each of the three RFPL genes encodes two exons giving rise to a putative RING-like motifs and B30-2 domains. RFPL2, or RNF79, is 378 amino acids long and is also highly expressed in prostate with lower expression in fetal kidney and fetal liver. As a result of alternative splicing, two isoforms of RFPL2 exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFPL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RFPL2
Gene Accession No. O75678
Gene Alias ret finger protein like 2; ret finger protein-like 2; RFPL2; RING finger protein 79; RNF79
Gene Symbols RFPL2
Host Species Rabbit
Immunogen Recombinant full length Human RFPL2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10739
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.