Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589934 Supplier Invitrogen™ Supplier No. PA589934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PMSLECGCAV CLKCINSLQK EPHGEDLLCC CSSMVSRKNK IRRNRQLERL ASHIKELEPK LKKILQMNPR MRKFQVDMTL DANTANNFLL ISDDLRSVRS G; Positive Samples: U-87MG, 22RV1, 293T, LO2, Rat brain, Rat testis.

RFPL1, RFPL2 and RFPL3 (ret finger protein-like 1, 2 and 3, respectively), exist as a cluster of genes mapping to human chromosome 22q12.2-q13.3, sharing 95%-96% identity. RFPL1, 2 and 3, are thought to contribute to neocortex organization and size in primates, and show high expression in fetal neocortex as well as embryonic stem-cell neurogenesis. Each of the three RFPL genes encodes two exons giving rise to a putative RING-like motifs and B30-2 domains. RFPL2, or RNF79, is 378 amino acids long and is also highly expressed in prostate with lower expression in fetal kidney and fetal liver. As a result of alternative splicing, two isoforms of RFPL2 exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFPL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RFPL2
Gene Accession No. O75678
Gene Alias ret finger protein like 2; ret finger protein-like 2; RFPL2; RING finger protein 79; RNF79
Gene Symbols RFPL2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human RFPL2 (NP_0065962).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10739
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.