Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFPL3 Antibody (OTI2G4), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273862MFV450
Description
RFPL3 Monoclonal antibody specifically detects RFPL3 in Human samples. It is validated for Flow CytometrySpecifications
RFPL3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI2G4 | |
Flow Cytometry | |
ret finger protein-like 3 | |
Full length human recombinant protein of human RFPL3(NP_006595) produced in HEK293T cell. | |
0.1 mL | |
Zinc Finger | |
10738 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction