Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFPL3 Antibody (OTI2G4), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273862PEATT594
Description
RFPL3 Monoclonal specifically detects RFPL3 in Human samples. It is validated for Flow Cytometry.Specifications
RFPL3 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
RFPL3 | |
Full length recombinant protein of human RFPL3 (NP_006595) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Flow Cytometry | |
OTI2G4 | |
Flow Cytometry | |
ret finger protein-like 3 | |
Mouse | |
Affinity Purified | |
Zinc Finger | |
10738.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction