Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFPL3 Monoclonal Antibody (OTI2A6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504733S


Description
RFPL3 Monoclonal antibody specifically detects RFPL3 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
RFPL3 | |
Monoclonal | |
1.04 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O75679 | |
Mouse | |
Affinity chromatography | |
RUO | |
10738 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI2A6 | |
Unconjugated | |
RFPL3 | |
ret finger protein like 3; ret finger protein-like 3; RFPL3 | |
Full length human recombinant protein of human RFPL3 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction