Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RFPL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567240 Supplier Invitrogen™ Supplier No. PA567240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEASSCPVCSD YLEKPMSLEC GCTVC Highest antigen sequence identity to the following orthologs - mouse 54%, rat 50%.

RFPL3 stimulates the activity of Human Immunodeficiency Virus 1/HIV-1 pre-integration complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFPL3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFPL3
Gene Accession No. O75679
Gene Alias ret finger protein like 3; ret finger protein-like 3; RFPL3
Gene Symbols RFPL3
Host Species Rabbit
Immunogen Recombinant Human RFPL3. Recombinant protein control fragment (Product #RP-105661).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10738
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.