Learn More
Invitrogen™ RFPL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588495
Description
Immunogen sequence: MAALFQEASS CPVCSDYLEK PMSLECGCTV CLKCINSLQK EPHGEDLLCC CCSMVSQRNK IRPNRQLERL VSHIKELEPK LKKILQMNPR MRKFQVDMTL DADTANNFLL ISDDLRSVRS GLITQNRQDL AERFDVSVCI LGSPRFTCGR HYWEVDVGTS TEWDLGVCRE SVHCKGKIQL TTELGFWTVS LRDGSRLSAS TVPLTFLLVD RKLQRVGIFL DMGMQNVSFF DAESGSHVYT FRSVSAEEPL RPFLAPSIPP NGDQGVLSIC PLMNSGTTDA PVRPGEAK; Positive Samples: U-87MG, HeLa.
RFPL3 stimulates the activity of Human Immunodeficiency Virus 1/HIV-1 pre-integration complex.
Specifications
RFPL3 | |
Polyclonal | |
Unconjugated | |
RFPL3 | |
ret finger protein like 3; ret finger protein-like 3; RFPL3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10738 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.98 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O75679 | |
RFPL3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-288 of human RFPL3 (NP_0065951). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.